The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031378.3 | NP_113555.1 | 1 - 2 | 269 | 84 |
GenBank mRNA | AK137662.1 | - | 1 - 2 | 270 | 84 |