Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps33a
Entrez Gene ID
77573
Gene Name
vacuolar protein sorting 33A (yeast)
Gene Aliases
3830421M04Rik, AI503300, AW048546, AW554476, bf
Chromosome Location
Chr.5: 123527304 - 123573043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029929.3 NP_084205.3 4 - 5 523 86
GenBank mRNA AF439858.1 - 4 - 5 507 86
AK019463.1 - 4 - 5 507 86
AK041982.1 - 4 - 5 519 86
AK140553.1 - 4 - 5 512 86
AK150282.1 - 4 - 5 515 86
AK150550.1 - 4 - 5 515 86
BC046417.1 - 4 - 5 514 86

View Full Product Details