The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033461.3 | NP_001028633.1 | 11 - 12 | 1883 | 82 |
NM_001289821.1 | NP_001276750.1 | 11 - 12 | 1862 | 82 | |
GenBank mRNA | AK133051.1 | - | 11 - 12 | 1884 | 82 |
AK167318.1 | - | 11 - 12 | 1861 | 82 | |
AK167729.1 | - | 11 - 12 | 1861 | 82 |