Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myrf
Entrez Gene ID
225908
Gene Name
myelin regulatory factor
Gene Aliases
6030439E18, Gm1804, Gm98, Mrf
Chromosome Location
Chr.19: 10208271 - 10240781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033481.1 NP_001028653.1 9 - 10 1532 84
GenBank mRNA AK141695.1 - 9 - 10 1533 84
BC157942.1 - 6 - 7 923 84

View Full Product Details