Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Micall2
Entrez Gene ID
231830
Gene Name
MICAL-like 2
Gene Aliases
A930021H16Rik, JRAB, MICAL-L2, mFLJ00139
Chromosome Location
Chr.5: 139706692 - 139736333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174850.3 NP_777275.2 17 - 17 3198 159
GenBank mRNA AK044547.1 - 17 - 18 67
AK151086.1 - 17 - 17 3197 159
AK164043.1 - 17 - 17 3169 159
AK165558.1 - 17 - 17 3048 159
BC043300.1 - 17 - 17 3012 159
BY700282.1 - 2 - 2 240 159

View Full Product Details