The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081462.2 | NP_001074931.1 | 14 - 15 | 1830 | 72 |
NM_001081463.2 | NP_001074932.1 | 14 - 15 | 1830 | 72 | |
NM_001081464.2 | NP_001074933.1 | 14 - 15 | 1830 | 72 | |
NM_001081465.1 | NP_001074934.1 | 14 - 15 | 1773 | 72 | |
NM_001081466.2 | NP_001074935.1 | 14 - 15 | 1830 | 72 | |
NM_001081467.2 | NP_001074936.1 | 14 - 15 | 1830 | 72 | |
NM_001081468.2 | NP_001074937.1 | 14 - 15 | 1830 | 72 | |
NM_001081469.2 | NP_001074938.1 | 14 - 15 | 1830 | 72 | |
NM_001081470.2 | NP_001074939.1 | 14 - 15 | 1830 | 72 | |
NM_001244936.1 | NP_001231865.1 | 14 - 15 | 1830 | 72 | |
NM_020331.3 | NP_065064.2 | 14 - 15 | 1830 | 72 | |
GenBank mRNA | AF247161.1 | - | 14 - 15 | 1690 | 72 |
AF257475.1 | - | 14 - 15 | 1806 | 72 | |
AF260133.1 | - | 14 - 15 | 1801 | 72 | |
AF343348.1 | - | 14 - 15 | 1698 | 72 | |
AF343349.1 | - | 14 - 15 | 1704 | 72 | |
AF343350.1 | - | 14 - 15 | 1700 | 72 | |
AF343351.1 | - | 14 - 15 | 1698 | 72 | |
AF497637.1 | - | 14 - 15 | 1690 | 72 | |
AF497638.1 | - | 14 - 15 | 1690 | 72 | |
AF497639.1 | - | 14 - 15 | 1690 | 72 | |
AF497640.1 | - | 14 - 15 | 1698 | 72 | |
AF497641.1 | - | 14 - 15 | 1704 | 72 | |
AF497642.1 | - | 14 - 15 | 1700 | 72 | |
AK078488.1 | - | 14 - 15 | 1698 | 72 | |
AK079487.1 | - | 14 - 15 | 1770 | 72 | |
AK147196.1 | - | 14 - 15 | 1813 | 72 | |
AY030287.1 | - | 14 - 15 | 1801 | 72 | |
AY030288.1 | - | 14 - 15 | 1801 | 72 | |
AY030289.1 | - | 14 - 15 | 1801 | 72 | |
BC050822.1 | - | 14 - 15 | 1825 | 72 | |
BC057091.1 | - | 14 - 15 | 1652 | 72 | |
BC058964.1 | - | 14 - 15 | 1652 | 72 | |
BC083150.1 | - | 14 - 15 | 1732 | 72 |