The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033572.2 | NP_291050.1 | 10 - 11 | 1362 | 74 |
GenBank mRNA | AF410456.1 | - | 10 - 11 | 1341 | 74 |
AK005040.1 | - | 10 - 11 | 1309 | 74 | |
AK017733.1 | - | 10 - 11 | 1361 | 74 | |
BC024714.1 | - | 10 - 11 | 1334 | 74 | |
BC055335.1 | - | 10 - 11 | 1360 | 74 |