The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114339.2 | NP_001107811.1 | 2 - 3 | 1289 | 128 |
NM_023792.2 | NP_076281.1 | 2 - 3 | 589 | 128 | |
GenBank mRNA | AF200357.1 | - | 2 - 3 | 589 | 128 |
AF347700.1 | - | 2 - 3 | 1287 | 128 | |
AK017345.1 | - | 2 - 3 | 1156 | 128 | |
AK050222.1 | - | 2 - 3 | 590 | 128 | |
BC023496.1 | - | 2 - 3 | 511 | 128 |