Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ermp1
Entrez Gene ID
226090
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815
Chromosome Location
Chr.19: 29609883 - 29648484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081213.1 NP_001074682.1 3 - 4 817 67
GenBank mRNA AK041066.1 - 3 - 4 788 67
AK050284.1 - 3 - 4 813 67
AK081674.1 - 2 - 3 334 67
AK129447.1 - 3 - 4 745 67
AK137206.1 - 3 - 4 773 67
BC086627.1 - 3 - 4 758 67
BC151181.1 - 3 - 4 776 67
BC151182.1 - 3 - 4 776 67
BC157914.1 - 3 - 4 776 67

View Full Product Details