Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ermp1
Entrez Gene ID
226090
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815
Chromosome Location
Chr.19: 29609883 - 29648484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081213.1 NP_001074682.1 4 - 5 920 64
GenBank mRNA AK041066.1 - 4 - 5 891 64
AK081674.1 - 3 - 4 437 64
AK129447.1 - 4 - 5 848 64
AK137206.1 - 4 - 5 876 64
BC086627.1 - 4 - 5 861 64
BC151181.1 - 4 - 5 879 64
BC151182.1 - 4 - 5 879 64
BC157914.1 - 4 - 5 879 64

View Full Product Details