Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ide
Entrez Gene ID
15925
Gene Name
insulin degrading enzyme
Gene Aliases
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location
Chr.19: 37268741 - 37334544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031156.3 NP_112419.3 15 - 16 2023 99
GenBank mRNA AJ278422.1 - 15 - 16 2043 99
AK004972.2 - 15 - 16 2025 99
AK128986.1 - 15 - 16 2040 99
BC041675.1 - 15 - 16 2055 99

View Full Product Details