Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brdt
Entrez Gene ID
114642
Gene Name
bromodomain, testis-specific
Gene Aliases
7420412D09Rik, Brd6, Fsrg3
Chromosome Location
Chr.5: 107331091 - 107387061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054054.2 NP_473395.2 14 - 15 2450 122

View Full Product Details