The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081107.1 | NP_001074576.1 | 1 - 2 | 310 | 79 |
GenBank mRNA | AK040335.1 | - | 1 - 2 | 294 | 79 |
AK050769.1 | - | 1 - 2 | 287 | 79 | |
AK052427.1 | - | 1 - 2 | 270 | 79 | |
BC082601.1 | - | 1 - 2 | 274 | 79 | |
BC109169.2 | - | 1 - 2 | 178 | 79 | |
BC109170.2 | - | 1 - 2 | 183 | 79 |