The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033581.1 | 3 - 4 | 326 | 85 | |
GenBank mRNA | AK004863.1 | - | 3 - 4 | 332 | 85 |
BY705068.1 | - | 3 - 4 | 327 | 85 |