The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122639.2 | NP_001116111.1 | 5 - 6 | 688 | 100 |
NM_198306.3 | NP_938048.2 | 10 - 11 | 2018 | 100 | |
NR_104449.1 | 5 - 6 | 725 | 100 | ||
GenBank mRNA | AK032568.1 | - | 10 - 11 | 2021 | 100 |
AK170381.1 | - | 5 - 6 | 711 | 100 | |
AK171529.1 | - | 5 - 6 | 671 | 100 | |
BC054061.1 | - | 3 - 4 | 323 | 100 |