Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp12
Entrez Gene ID
231866
Gene Name
zinc finger protein 12
Gene Aliases
C530015C18, Krox-7, Zfp-12, Znf12
Chromosome Location
Chr.5: 143235163 - 143248834 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289589.1 NP_001276518.1 2 - 3 284 63
NM_001289590.1 NP_001276519.1 2 - 3 284 63
NM_177681.4 NP_808349.1 2 - 3 284 63
GenBank mRNA AK049650.1 - 2 - 3 240 63
BB612291.1 - 2 - 3 284 63
BC053080.1 - 2 - 3 193 63
BC059836.1 - 2 - 3 195 63

View Full Product Details