Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc29a4
Entrez Gene ID
243328
Gene Name
solute carrier family 29 (nucleoside transporters), member 4
Gene Aliases
ENT4, mPMAT
Chromosome Location
Chr.5: 142692476 - 142722490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146257.2 NP_666369.1 3 - 4 505 74
GenBank mRNA BC025599.1 - 3 - 4 449 74

View Full Product Details