Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
50765
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr.5: 137569822 - 137588081 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289507.1 NP_001276436.1 17 - 18 2326 67
NM_001289509.1 NP_001276438.1 19 - 20 2345 67
NM_001289511.1 NP_001276440.1 18 - 19 2203 67
NM_015799.4 NP_056614.3 19 - 20 2285 67
GenBank mRNA AF207741.1 - 17 - 18 2160 67
AF222895.1 - 18 - 19 2203 67
AK004848.2 - 19 - 20 2261 67
AK004965.1 - 19 - 20 2316 67
AK155526.1 - 18 - 19 2422 67
AK165320.1 - 17 - 18 2153 67
BC013654.1 - 17 - 18 2326 67

View Full Product Details