Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbas
Entrez Gene ID
14467
Gene Name
glioblastoma amplified sequence
Gene Aliases
AV006093, Nipsnap2
Chromosome Location
Chr.5: 129725044 - 129758327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008095.4 NP_032121.3 8 - 9 771 110
GenBank mRNA AK170336.1 - 8 - 9 717 110
AK183347.1 - 1 - 2 93 110
BC055893.1 - 8 - 9 720 110

View Full Product Details