Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp6v0a2
Entrez Gene ID
21871
Gene Name
ATPase, H+ transporting, lysosomal V0 subunit A2
Gene Aliases
8430408C20Rik, AI385560, ATP6a2, AW489264, Atp6n1d, Atp6n2, C76904, ISF, J6B7, SHIF, Stv1, TJ6M, TJ6s, Tj6, V-ATPase 116 kDa, V-ATPase a2
Chromosome Location
Chr.5: 124629052 - 124646816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011596.5 NP_035726.2 - 419 61
GenBank mRNA AB022323.1 - - 385 61
AF218252.1 - - 404 61
AK155055.1 - - 396 61
AK170122.1 - - 406 61
AK182205.1 - - 47 61
BC018369.1 - - 372 61
BC108991.1 - - 316 61
BC108992.1 - - 316 61
BC112905.1 - - 379 61
M31226.1 - - 315 61
X55184.1 - - 316 61

View Full Product Details