The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144509.2 | NP_653092.1 | 2 - 3 | 779 | 84 |
GenBank mRNA | AB035383.1 | - | 2 - 3 | 779 | 84 |
AK133934.1 | - | 2 - 3 | 595 | 84 | |
BC063748.1 | - | 2 - 3 | 746 | 84 |