Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psrc1
Entrez Gene ID
56742
Gene Name
proline/serine-rich coiled-coil 1
Gene Aliases
5430413I02Rik, AL033312, Dda3
Chromosome Location
Chr.3: 108383804 - 108388231 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190161.1 NP_001177090.1 2 - 3 141 61
NM_019976.3 NP_064360.1 2 - 3 138 61
GenBank mRNA AF156598.1 - 2 - 3 133 61
AK011197.1 - 2 - 3 113 61
AK017302.1 - 2 - 3 80 61
AK041835.1 - 2 - 3 88 61
AK167415.1 - 2 - 3 84 61
AK178652.1 - 2 - 3 133 61
BC019165.1 - 2 - 3 96 61
BY080522.1 - 2 - 3 138 61
BY231608.1 - 2 - 3 141 61

View Full Product Details