Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc60
Entrez Gene ID
269693
Gene Name
coiled-coil domain containing 60
Gene Aliases
AU018954, C130098D09
Chromosome Location
Chr.5: 116125581 - 116288998 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177759.3 NP_808427.1 7 - 8 1315 62
GenBank mRNA AK082047.1 - 7 - 8 1316 62
BC150892.1 - 7 - 8 1105 62
BC172040.1 - 7 - 8 1103 62

View Full Product Details