Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd1
Entrez Gene ID
107765
Gene Name
ankyrin repeat domain 1 (cardiac muscle)
Gene Aliases
Alrp, CARP, Crap, MARP1
Chromosome Location
Chr.19: 36111965 - 36119844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013468.3 NP_038496.2 4 - 5 512 103
GenBank mRNA AK009655.1 - 4 - 5 505 103
AK009959.1 - 4 - 5 511 103
AK145944.1 - 4 - 5 553 103
AK146471.1 - 4 - 5 509 103
AK146627.1 - 4 - 5 511 103
AK146940.1 - 4 - 5 509 103
BC037138.1 - 4 - 5 512 103

View Full Product Details