Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prune2
Entrez Gene ID
353211
Gene Name
prune homolog 2 (Drosophila)
Gene Aliases
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367
Chromosome Location
Chr.19: 16955670 - 17223958 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK020727.1 - 6 - 7 890 87
AK165913.1 - 6 - 7 890 87

View Full Product Details