Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpine1
Entrez Gene ID
18787
Gene Name
serine (or cysteine) peptidase inhibitor, clade E, member 1
Gene Aliases
PAI-1, PAI1, Planh1
Chromosome Location
Chr.5: 137061504 - 137072272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008871.2 NP_032897.2 1 - 2 144 59
GenBank mRNA AK040662.1 - 1 - 2 141 59
BC054091.1 - 1 - 2 141 59
M33960.1 - 1 - 2 130 59

View Full Product Details