Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpine1
Entrez Gene ID
18787
Gene Name
serine (or cysteine) peptidase inhibitor, clade E, member 1
Gene Aliases
PAI-1, PAI1, Planh1
Chromosome Location
Chr.5: 137061504 - 137072272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008871.2 NP_032897.2 6 - 7 1140 64
GenBank mRNA BC054091.1 - 6 - 7 1137 64
GQ872458.1 - 5 - 6 995 64
GQ872459.1 - 5 - 6 995 64
M33960.1 - 6 - 7 1126 64

View Full Product Details