Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oplah
Entrez Gene ID
75475
Gene Name
5-oxoprolinase (ATP-hydrolysing)
Gene Aliases
1700010G02Rik
Chromosome Location
Chr.15: 76296603 - 76331101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153122.2 NP_694762.1 10 - 11 1494 78
GenBank mRNA AK158391.1 - 10 - 11 1536 78
BC034522.1 - 10 - 11 1494 78

View Full Product Details