The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033312.3 | NP_001028484.2 | 4 - 5 | 2102 | 88 |
GenBank mRNA | AK049302.1 | - | 4 - 5 | 2056 | 88 |
AK144757.1 | - | 4 - 5 | 2104 | 88 | |
AK154354.1 | - | 4 - 5 | 2082 | 88 |