Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxh1
Entrez Gene ID
14106
Gene Name
forkhead box H1
Gene Aliases
Fast1, Fast2, fast-1, fast-2
Chromosome Location
Chr.15: 76667829 - 76670317 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007989.4 NP_032015.1 2 - 3 453 84
GenBank mRNA AB221574.1 - 2 - 3 381 84
AF069303.1 - 2 - 3 379 84
AF079514.1 - 2 - 3 376 84
AF110506.1 - 2 - 3 376 84
AF177770.1 - 2 - 3 376 84
BC119156.1 - 2 - 3 453 84
BC137781.1 - 2 - 3 453 84

View Full Product Details