The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152923.2 | NP_690887.2 | 7 - 8 | 1143 | 98 |
GenBank mRNA | AY118171.1 | - | 7 - 8 | 1143 | 98 |
BC116306.1 | - | 7 - 8 | 1134 | 98 |