The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146208.2 | NP_666320.1 | 8 - 9 | 1611 | 84 |
GenBank mRNA | AB072931.1 | - | 8 - 9 | 1478 | 84 |
BC024921.1 | - | 8 - 9 | 1470 | 84 | |
BC034753.1 | - | 8 - 9 | 1554 | 84 |