The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011258.2 | NP_035388.2 | 1 - 2 | 115 | 127 |
GenBank mRNA | AK132028.1 | - | 1 - 2 | 95 | 127 |
AK136937.1 | - | 1 - 2 | 94 | 127 | |
AK146164.1 | - | 1 - 2 | 110 | 127 | |
AK146314.1 | - | 1 - 2 | 110 | 127 | |
BC085173.1 | - | 1 - 2 | 92 | 127 | |
BC085174.1 | - | 1 - 2 | 84 | 127 | |
BY714128.1 | - | 1 - 2 | 143 | 127 | |
U36441.1 | - | 1 - 2 | 92 | 127 | |
X72711.1 | - | 1 - 2 | 81 | 127 |