Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube3c
Entrez Gene ID
100763
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
AI853514, mKIAA0010
Chromosome Location
Chr.5: 29569242 - 29676077 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133907.3 NP_598668.1 3 - 4 468 111
GenBank mRNA AK029973.1 - 3 - 4 278 111
AK045211.1 - 3 - 4 265 111
AK046056.1 - 3 - 4 469 111
AK122187.1 - 3 - 4 403 111
AK140491.1 - 3 - 4 225 111
BC120731.1 - 3 - 4 215 111
BC137626.1 - 3 - 4 215 111

View Full Product Details