The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205132.1 | NP_001192061.1 | 9 - 10 | 1337 | 126 |
NM_001205133.1 | NP_001192062.1 | 9 - 10 | 1337 | 126 | |
NM_009535.3 | NP_033561.1 | 9 - 10 | 1799 | 126 | |
GenBank mRNA | AK035612.1 | - | 9 - 10 | 1786 | 126 |
AK052478.1 | - | 9 - 10 | 1289 | 126 | |
AK167420.1 | - | 9 - 10 | 1474 | 126 | |
BC010594.1 | - | 9 - 10 | 1173 | 126 | |
X67677.1 | - | 9 - 10 | 1726 | 126 |