The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025700.2 | NP_079976.1 | 3 - 4 | 417 | 101 |
GenBank mRNA | AK014332.1 | - | 3 - 4 | 342 | 101 |
BC030869.1 | - | 3 - 4 | 379 | 101 | |
BC052762.1 | - | 3 - 4 | 417 | 101 |