Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrg1
Entrez Gene ID
211323
Gene Name
neuregulin 1
Gene Aliases
6030402G23Rik, ARIA, D230005F13Rik, GGF, GGFII, HRG, HRGalpha, Hgl, NDF, SMDF
Chromosome Location
Chr.8: 31807452 - 32890583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178591.2 NP_848706.2 1 - 2 667 104
GenBank mRNA AK048708.1 - 1 - 2 849 104
AK141037.1 - 1 - 2 855 104
AY648975.1 - 1 - 2 667 104
BC151113.1 - 1 - 2 751 104

View Full Product Details