Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkx
Entrez Gene ID
19108
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1, Pkare
Chromosome Location
Chr.X: 77760972 - 77796288 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016979.1 NP_058675.1 8 - 9 1353 159
GenBank mRNA AK034681.1 - 8 - 9 1672 159
AK037141.1 - 8 - 9 1347 159
AK081548.1 - 8 - 9 1333 159
AK139510.1 - 8 - 9 1361 159
AK149937.1 - 8 - 9 1331 159
AK150588.1 - 8 - 9 1315 159
AK152166.1 - 8 - 9 1149 159
AK154447.1 - 8 - 9 1333 159
AK169322.1 - 8 - 9 1304 159
BC006875.1 - 8 - 9 1353 159

View Full Product Details