The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161824.1 | NP_001155296.1 | - | ||
NM_013604.2 | NP_038632.2 | - | |||
GenBank mRNA | AK086135.1 | - | - | ||
AK159316.1 | - | - | |||
AK163982.1 | - | - | |||
AK165923.1 | - | - | |||
AX775987.1 | - | - | |||
BB662550.1 | - | - | |||
BC023071.1 | - | - | |||
L36962.1 | - | - |