Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtx1
Entrez Gene ID
17827
Gene Name
metaxin 1
Gene Aliases
Gcap6, Mtx
Chromosome Location
Chr.3: 89209081 - 89214347 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161824.1 NP_001155296.1 7 - 8 1184 90
NM_013604.2 NP_038632.2 7 - 8 1187 90
GenBank mRNA AK086135.1 - 7 - 8 780 90
AK159316.1 - 6 - 7 874 90
AK163982.1 - 7 - 8 755 90
AK165547.1 - 7 - 8 762 90
AK165923.1 - 7 - 8 760 90
AX775987.1 - 7 - 8 759 90
BC023071.1 - 7 - 8 758 90
L36962.1 - 7 - 8 757 90

View Full Product Details