Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sult1b1
Entrez Gene ID
56362
Gene Name
sulfotransferase family 1B, member 1
Gene Aliases
ST1B1, St2b2
Chromosome Location
Chr.5: 87513339 - 87538195 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019878.4 NP_063931.1 6 - 7 706 102
GenBank mRNA AF022894.1 - 6 - 7 725 102
AK033498.1 - 6 - 7 709 102
AK087595.1 - 6 - 7 742 102
BC024361.1 - 5 - 6 665 102
U92076.1 - 6 - 7 651 102

View Full Product Details