The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010775.2 | NP_034905.1 | 3 - 4 | 394 | 74 |
GenBank mRNA | AK034788.1 | - | 3 - 4 | 394 | 74 |
BC012245.1 | - | 3 - 4 | 356 | 74 | |
BC021762.1 | - | 3 - 4 | 348 | 74 | |
D11441.1 | - | 3 - 4 | 338 | 74 | |
S42292.1 | - | 3 - 4 | 394 | 74 |