The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080813.2 | NP_001074282.1 | 1 - 2 | 441 | 55 |
NM_029423.2 | NP_083699.2 | 1 - 2 | 441 | 55 | |
GenBank mRNA | AK014696.1 | - | 1 - 2 | 423 | 55 |
AK030769.1 | - | 1 - 2 | 444 | 55 | |
AK086797.1 | - | 1 - 2 | 439 | 55 | |
AK151024.1 | - | 1 - 2 | 442 | 55 | |
BB619642.1 | - | 1 - 2 | 442 | 55 | |
BC125400.1 | - | 1 - 2 | 426 | 55 | |
BC132094.1 | - | 1 - 2 | 426 | 55 |