Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ss18l1
Entrez Gene ID
269397
Gene Name
synovial sarcoma translocation gene on chromosome 18-like 1
Gene Aliases
A230053O16Rik, CREST
Chromosome Location
Chr.2: 180042483 - 180070201 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178750.5 NP_848865.4 4 - 5 691 64
GenBank mRNA AK038669.1 - 4 - 5 386 64
AK054560.1 - 4 - 5 406 64
AK133944.1 - 4 - 5 628 64
AK173004.1 - 4 - 5 444 64
BC053087.1 - 4 - 5 515 64
BC054730.1 - 4 - 5 665 64

View Full Product Details