Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrnad1
Entrez Gene ID
229503
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 87922601 - 87930588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC012224.1 - 2 - 3 403 116
BC023814.1 - 1 - 2 351 116

View Full Product Details