Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpy19l4
Entrez Gene ID
381510
Gene Name
dpy-19-like 4 (C. elegans)
Gene Aliases
Gm1023, Narg3
Chromosome Location
Chr.4: 11261315 - 11322135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081201.1 NP_001074670.1 16 - 17 1834 155
GenBank mRNA AK144417.1 - 16 - 17 1786 155
BC147500.1 - 16 - 17 1790 155
BC157932.1 - 16 - 17 1790 155
BC157988.1 - 16 - 17 1790 155

View Full Product Details