Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Setd4
Entrez Gene ID
224440
Gene Name
SET domain containing 4
Gene Aliases
AY037804, C21orf18, ORF21
Chromosome Location
Chr.16: 93583457 - 93604833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC029101.1 - 7 - 8 1657 87

View Full Product Details