Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddhd2
Entrez Gene ID
72108
Gene Name
DDHD domain containing 2
Gene Aliases
2010305K11Rik, SAMWD1, mKIAA0725
Chromosome Location
Chr.8: 25725324 - 25754590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028102.1 NP_082378.1 15 - 16 1962 83
GenBank mRNA AK173010.1 - 11 - 12 2392 83
BC046229.1 - 15 - 16 1882 83
BC118962.2 - 13 - 14 1955 83

View Full Product Details