Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chd7
Entrez Gene ID
320790
Gene Name
chromodomain helicase DNA binding protein 7
Gene Aliases
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis
Chromosome Location
Chr.4: 8690406 - 8868449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277149.1 NP_001264078.1 28 - 29 6189 88

View Full Product Details