The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271725.1 | NP_001258654.1 | 2 - 3 | 336 | 57 |
NM_001271726.1 | NP_001258655.1 | 2 - 3 | 372 | 57 | |
NM_001271727.1 | NP_001258656.1 | 2 - 3 | 641 | 57 | |
NM_001271728.1 | NP_001258657.1 | 2 - 3 | 439 | 57 | |
NM_030706.3 | NP_109631.2 | 2 - 3 | 435 | 57 | |
GenBank mRNA | AB043550.1 | - | 2 - 3 | 419 | 57 |
AF220017.1 | - | 2 - 3 | 284 | 57 | |
AK039741.1 | - | 2 - 3 | 433 | 57 | |
AK045410.1 | - | 2 - 3 | 432 | 57 | |
AK046887.1 | - | 2 - 3 | 420 | 57 | |
AK079415.1 | - | 2 - 3 | 586 | 57 | |
AK141419.1 | - | 2 - 3 | 282 | 57 | |
AK147275.1 | - | 2 - 3 | 549 | 57 | |
AK147395.1 | - | 2 - 3 | 430 | 57 | |
AK164350.1 | - | 1 - 2 | 138 | 57 | |
AK220343.1 | - | 2 - 3 | 640 | 57 | |
BC058668.1 | - | 2 - 3 | 403 | 57 | |
BC058961.1 | - | 2 - 3 | 439 | 57 | |
CD802800.1 | - | 2 - 3 | 298 | 57 | |
CD803806.1 | - | 2 - 3 | 435 | 57 | |
CJ100776.1 | - | 2 - 3 | 337 | 57 |